Learn More
DWK Life Sciences Kimble™ Amber Plain Disposable Borosilicate Glass Tubes
Produced from amber borosilicate glass to give superior protection for light sensitive applications.
Supplier:  DWK Life Sciences 7350013100A
 View more versions of this product
Catalog No. 13678256
Only null left
Description
Description
  • Amber 51 expansion borosilicate glass offers excellent chemical resistance against sodium leaching, a factor common in soda lime glass products
  • Tubes are packed in convenient trays for ease of use on bench tops and in drawers
  • Trays are shrink-wrapped in plastic film, with tubes remaining in constant alignment for protection against in-transit breakage and contamination 
  • No marking spot
  • Designed from ASTM Specification E890
  • Manufactured from amber 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Specifications
Chemical Identifiers
Amber
Single-use
None
Glass
Light sensitive applications
1000/Cs; Shrink-wrapped trays
Specifications
Amber
100 mm
Single-use
None
13 mm
10 mL
Glass
Light sensitive applications
1000/Cs; Shrink-wrapped trays
Disposable Culture Tube
Product Suggestions
Product Suggestions