Learn More
DWK LIFE SCIENCES™ KIMBLE™ Open Top Linerless Polypropylene Screw Thread Caps
Catalog No. 03340124D
Change view
Click to view available options
Color:
Black
Catalog No. Color
03340124D Black
1 options
Catalog No. 03340124D Supplier DWK LIFE SCIENCES™ Supplier No. 7380624400
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
KIMBLE™ Open Top Linerless Polypropylene Screw Thread Caps are economical, one-piece linerless closures constructed of durable polypropylene. Autoclavable and available in multiple GPI finishes for reliable, repeated use across laboratory applications.
  • Constructed of durable polypropylene for chemical resistance and durability
  • Economical, one-piece linerless construction
  • Open-top design for easy access
  • Autoclavable for repeated sterilization cycles
  • Available in multiple GPI finish sizes to suit a range of bottle and vial threads
Specifications
Diameter (Metric) Opening 1.4 cm
Color Black
Diameter (Metric) Outer 24 mm
Material Polypropylene
Pre-slit No
Sterility Non-sterile
Quantity 1000/Cs.
Closure Size 24-400
Autoclavable Autoclavable
Product Type Screw Cap