missing translation for 'onlineSavingsMsg'
Learn More

DWK LIFE SCIENCES™ KIMBLE™ Black with Cone-Shaped Liner Phenolic Caps/Closures

Catalog No. 03340210
Change view
Click to view available options
Closure Size:
13-425
18-400
22-400
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size
03340210 22-400
03340206 13-425
03340208 18-400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03340210 Supplier DWK LIFE SCIENCES™ Supplier No. 7380922400

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

KIMBLE™ Black with Cone-Shaped Liner Phenolic Caps/Closures provide excellent sample storage and re-sealing properties. Made from specially formulated phenolic resin, these caps feature LDPE cone-shaped liners that create a two-part seal for reliable closure and superior torque retention.

  • Specially formulated phenolic resin withstands the effects of repeated autoclaving
  • Cone-shaped LDPE liner provides a two-part seal: cap seals on the container rim, liner seals at the inner diameter of the container opening
  • Resistant to stress cracking
  • Designed for superior torque retention
  • Excellent for sample storage and re-sealing

Specifications

Material (Liner) Polyethylene
Color Black
Diameter (Metric) Outer 22 mm
Material Phenolic
Pre-slit No
Sterility Non-sterile
For Use With (Application) Excellent sample storage and re-sealing properties
Quantity 144/Cs.
Closure Size 22-400
Autoclavable Not Autoclavable
Product Type Screw Cap
Show More Show Less