missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Aluminum Seal Caps

Catalog No. 03341602
Change view
Click to view available options
Color:
Natural
Red
No. per Case:
100/Cs.
1000/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. per Case
03341602 Red 1000/Cs.
03341601 Natural 100/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03341602 Supplier DWK Life Sciences Supplier No. N7382611

With PTFE-faced rubber septa

  • Moderate to good resealability
  • One piece

  • PTFE-faced red rubber septa
  • For general GC applications
  • Moderate resealability
  • Not recommended for multiple injections
  • PTFE-faced natural rubber septa
  • Good resealability
  • Multiple injections

Specifications

Color Red
For Use With (Application) Multiple injection
Material Aluminum
No. per Case 1000/Cs.
Liner Material PTFE/Rubber
Neck Finish 11