Learn More
DWK Life Sciences Kimble™ Aluminum Heating Blocks for Kimble-Chase Kontes™ Microflex™ Microscale Kits
Use with Mfr. No. K309180-0290 Metallic Thermometer.
Supplier:  DWK Life Sciences 7202000001
 View more versions of this product
Catalog No. K7202000001
Only null left
Description
Description
  • Ideal for use in place of a sand bath in microscale experiments
  • Excellent heat transfer
  • Does not interfere with magnetic stirring
Specifications
Chemical Identifiers
Aluminum
Ideal for use in place of a sand bath in microscale experiments
3 in.
3 in.
0.75 in.
Heating Block
3 x 3 x 0.75 in. (76.2 x 76.2 x 19.05 mm)
76.2 mm
76.2 mm
19.05 mm
Specifications
Standard Taper Size 14/10 Kit
Heating Block
3 x 3 x 0.75 in. (76.2 x 76.2 x 19.05 mm)
76.2 mm
76.2 mm
19.05 mm
Aluminum
Ideal for use in place of a sand bath in microscale experiments
3 in.
3 in.
0.75 in.
Product Suggestions
Product Suggestions