Learn More
Kimberly-Clark Professional™ WypAll™ X60 Wipers
Fast absorption and outstanding tear resistance
Supplier:  Kimberly-Clark Professional™ 35421
 View more versions of this product
Catalog No. 19014876
Only null left
Description
Description
Fast absorption and outstanding tear resistance
  • Made from Hydroknit™, an advanced wiper material manufactured using jets of water to bond soft, absorbent paper to extra-strong, nonwoven fabric
  • Reinforced for extra scrubbing and cleaning power
  • No adhesives or binders—soft enough for face and hands
Specifications
Chemical Identifiers
Cleaning Cloth
Non-woven Polyproylene Cellulose
Specifications
Cleaning Cloth
34 cm
50.29 cm
White
13.4 x 19.8 in.
13.4 in.
13.4 in.
Non-woven Polyproylene Cellulose
Single-use
General purpose cleaning and wiping of surfaces, equipment, and hands and faces
Product Suggestions
Product Suggestions