Learn More
Ansell Kimtech™ A8 Certified Lab Coats with Knit Cuffs + Extra Protection
A8 certified lab coats with extra protection are made for environments where low to moderate exposure to chemicals or fluids is a risk.
Supplier:  Ansell 10045
 View more versions of this product
Catalog No. 18-022834
Only null left
Explore available promotions
Description
Description
  • 3-layer SMS nonwoven fabric
  • Resistance to blood strike-through at various levels of exposure
  • Mid-calf length protective coat has knit cuffs and a traditional collar
  • Three reinforced pockets, six snaps and a back vent for increased comfort and ventilation
  • Neck loop makes it convenient to hang
  • For use when there is potential for low to moderate exposure to chemicals or fluids
Specifications
Specifications
General Purpose Lab Coat
Unisex
Blue
Knit
Non-sterile
3
Yes
30 to 34 in.
Small
SMS
Snaps
Single-use
Knee-length
Traditional
10045
Hanging Neck Loop
Product Suggestions
Product Suggestions