Learn More
Ansell Kimtech™ A8 Certified Lab Coats with Knit Cuffs + Extra Protection
GREENER_CHOICE
Catalog No. 18022831
Change view
Click to view available options
Color:
Blue
White
Size:
Small
Medium
Large
X-Large
2X-Large
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
18022831 White Large
18022829 White Small
18022830 White Medium
18022833 White 2X-Large
18022835 Blue Medium
18022836 Blue Large
18022837 Blue X-Large
18022832 White X-Large
18022838 Blue 2X-Large
18022834 Blue Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 18022831 Supplier Ansell Supplier No. ASL10042
Edge
A8 certified lab coats with extra protection are made for environments where low to moderate exposure to chemicals or fluids is a risk.
  • 3-layer SMS nonwoven fabric
  • Resistance to blood strike-through at various levels of exposure
  • Mid-calf length protective coat has knit cuffs and a traditional collar
  • Three reinforced pockets, six snaps and a back vent for increased comfort and ventilation
  • Neck loop makes it convenient to hang
  • For use when there is potential for low to moderate exposure to chemicals or fluids
Specifications
Product Type General Purpose Lab Coat
Size Large
Gender Unisex
Material SMS
Color White
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
Style Knee-length
No. of Pockets 3
Collar Type Traditional
Latex Free Yes
Model 10042
Size (English) Chest 42 to 46 in.
Includes Hanging Neck Loop
Show More Show Less