missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell X-Tra™ Traction Shoe Covers
Description
- Light blue, three-layer SMS fabric
- Fully elasticized
- Adhesive strips for traction on bottom
Specifications
Specifications
| Size | Medium/Large |
| Material | SMS |
| Coating | Polyethylene |
| Color | Blue |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Closure Type | Elastic Ankle |
| Sole Type | Traction Strips |
| Model | 69252 |
| Includes | Traction |
| Show More |