Learn More
Ansell Smith & Wesson™ Magnum 3G™ Eyewear
Ergonomically dimensioned three-point fit for comfort
$171.95 - $171.95
| Catalog Number | Mfr. No. | Size | Frame Color | Lens Tint | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Frame Color | Lens Tint | Price | Quantity | |||
|
19-150-2000
|
Ansell
19961 |
Full-sized | Platinum | Clear |
Case of 12 Pair for $171.95
|
|
|||
Description
Ergonomically dimensioned three-point fit for comfort
- Built-in hinge stops help to eliminate scratches
- Self-adjusting ergonomic rubber nose pad with rubber fingers
- Mini-Magnum™ designed to fit small faces
- Provides 99.9% UV protection
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
| Meets ANSI Z87.1-2003 standards. |