missing translation for 'onlineSavingsMsg'
Learn More

Ansell KleenGuard™ G10 Gray Nitrile Gloves

Catalog No. 19169970A
Change view
Click to view available options
Size:
X-Small
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19169970A X-Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19169970A Supplier Ansell Supplier No. 97820

KleenGuard™ G10 gloves provide durability and strength for light-duty tasks. Ideal for industrial, manufacturing and food processing applications.

  • Powder free, nitrile
  • Textured fingertips provide enhanced grip and dexterity
  • Ambidextrous
  • Beaded cuff
  • Puncture-resistant formulation delivers barrier protection against chemical splash, dirt, grease and oil
  • Ideal for cleaning and maintenance, manufacturing, food processing, electronic assembly and painting applications

Specifications

Certifications/Compliance FDA, CFIA approved
Applications Industrial
AQL 1.5
Color Gray
Exterior Finish Partially Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size X-Small
Thickness (English) 4 mil
Thickness (Metric) 0.10 mm
Model 97820
Length (English) 9.5 in.
Length (Metric) 24 cm
Show More Show Less