missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kimberly-Clark Professional™ KleenGuard™ A70 Chemical Spray Coveralls
Reflex™ design offers 12% more chest room and 6% longer body length
Manufacturer: Kimberly-Clark Professional™ KC00683
Description
Coveralls provide protection for hazardous waste remediation, environmental cleanup, petrochemical and oil refining, acid and caustic handling, biohazard cleanup, tank cleaning and pharmaceutical manufacturing.
- Feature bound seams
- Storm flap provides a seamless chest area
- Water-resistant zipper and wick-away lining for enhanced comfort
- Yellow
Compliance
Meet ASTM™ F 903 penetration test standards.
Specifications
Elastic | |
Polyethylene, Spunbond Polyproylene Fabric | |
Yellow | |
Zipper Front, Storm Flap, Elastic Wrists, Ankles, Hood and Boots | |
Hooded | |
Bound | |
1.5 mil |
Front zipper with storm flap | |
ASTM F 903, NFPA 99 | |
Elastic | |
Hazardous waste remediation, environmental cleanup, petrochemical refining, Acid/Caustic handling, biohazard cleanup, tank cleaning, pharmaceutical manufacturing | |
Chemical | |
Large |