missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell KleenGuard™ A60 Bloodborne Pathogen and Chemical Splash Protection Coveralls
Provide exceptional protection against nonhazardous liquids, dry particulates, and bloodborne pathogens, even under pressure
$181.63 - $181.63
Specifications
| Material | Fabric |
|---|---|
| Color | Denim Blue |
| Resistance | Bloodborne Pathogens, Chemicals |
| Certifications/Compliance | ASTM F1670, 1671 |
| For Use With (Application) | Animal research, emergency medical response, crime scene cleanup, embalming/forensics, tank cleaning, Fiberglass manufacturing, liquid handling, food processing |
| Catalog Number | Mfr. No. | Closure Type | Description | Size | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Closure Type | Description | Size | Price | Quantity | |||
|
19-057-402F
|
Ansell
ASL45237 |
Front Zipper | Seamless zipper front, open wrist and ankle, Reflex™ design; 3-ply Fabric construction middle layer of Microporous film | 4X-Large |
Case of 20 Each for $181.63
|
|
|||
Description
Breathable fabric is strong, comfortable, and abrasion-resistant, with a three-layer construction designed to allow heat and sweat vapor to escape.
- Patented Reflex™ Design is 7.5 times less likely to rip out than ANSI™ minimum standards
- Feature 12% more chest room and 6% longer body length
- Provide excellent front and strikethrough protection with a seamless front and concealed 28 in. (71cm) zipper
- Used in many industries and applications, including emergency medical, liquid handling, forensics, nuclear facilities and more
- Denim blue
Compliance
Meet ASTM F1670/1671 specifications for blood, bodily fluids and bloodborne pathogens.
Specifications
| Fabric | |
| Bloodborne Pathogens, Chemicals | |
| Animal research, emergency medical response, crime scene cleanup, embalming/forensics, tank cleaning, Fiberglass manufacturing, liquid handling, food processing |
| Denim Blue | |
| ASTM F1670, 1671 |