Learn More
Ansell KleenGuard™ A40 Liquid and Particle Protection Lab Coats
GREENER_CHOICE
Catalog No. p-2457790
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
No. of Pockets:
2
None
Cuff Style:
Elastic
Open
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size No. of Pockets Cuff Style
191202403C Large None Elastic
191202403D X-Large None Elastic
191202403B Medium None Elastic
191202403E 2X-Large None Elastic
191202403F 3X-Large None Elastic
191202404B Medium 2 Open
191202404C Large 2 Open
191202404E 2X-Large 2 Open
191202404F 3X-Large 2 Open
191202404D X-Large 2 Open
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 191202403C Supplier Ansell Supplier No. 44443
Only null left
Microporous lab coats provide protection against debris, non-hazardous liquid chemicals, and dry particulates.
Suited for use in fiberglass manufacturing, tank cleaning, utilities, liquid handling, pressure washing, maintenance and general manufacturing
  • Snap front closure
  • Elastic or open wrist styles
  • Available with or without pockets
  • Breathable, microporous film laminate
  • Low in lint
  • Traditional collar
  • Passes NFPA 99 criteria for antistatic materials
Compliance
Meets ASTM F1670 testing for penetration of blood and bodily fluids
Specifications
Product Type General Purpose Lab Coat
Size Large
Gender Unisex
Material Microporous Film
Color White
Closure Type Snaps
Cuff Style Elastic
Sterility Non-sterile
Style Knee-length
No. of Pockets None
Description No pockets and elastic wrists
Collar Type Traditional
Model 44443
Certifications/Compliance NFPA 99, ASTM F1670, ANSI ISEA 101-1996
For Use With (Application) Fiberglass Manufacturing, Tank Cleaning, Utilities, Liquid Handling, Pressure Washing, Maintenance, General Manufacturing
Show More Show Less