Learn More
Ansell KleenGuard™ A40 Liquid and Particle Protection Coveralls
Protective coveralls are made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals, and dry particulates.
$173.14 - $270.44
Specifications
Gender Unisex
Material Microporous Film Laminate
Color White
Resistance Particle, Liquid
View More Specs
Products 16
Catalog Number Mfr. No. Size Style Price Quantity  
Catalog Number Mfr. No. Size Style Price Quantity  
19-120-2398E
View Documents
Ansell
44305
2X-Large Open Wrists and Ankles N/A
Only null left
19-120-2398G
View Documents
Ansell
44307
4X-Large Open Wrists and Ankles N/A
Only null left
19-120-2398B
View Documents
Ansell
44302
Medium Open Wrists and Ankles N/A
Only null left
19-120-2398D
View Documents
Ansell
44304
X-Large Open Wrists and Ankles N/A
Only null left
19-120-2398C
View Documents
Ansell
44303
Large Open Wrists and Ankles N/A
Only null left
19-120-2398F
View Documents
Ansell
44306
3X-Large Open Wrists and Ankles N/A
Only null left
19-120-2400B
View Documents
Ansell
44322
Medium Elastic Wrists, Ankles with Hood N/A
Only null left
19-120-2400C
View Documents
Ansell
44323
Large Elastic Wrists, Ankles with Hood N/A
Only null left
19-120-2400D
View Documents
Ansell
44324
X-Large Elastic Wrists, Ankles with Hood N/A
Only null left
19-120-2400F
View Documents
Ansell
44326
3X-Large Elastic Wrists, Ankles with Hood N/A
Only null left
19-120-2400G
View Documents
Ansell
44327
4X-Large Elastic Wrists, Ankles with Hood N/A
Only null left
19-120-2401B
View Documents
Ansell
44332
Medium Elastic Wrists, Ankles with Hood and Boots N/A
Only null left
19-120-2401G
View Documents
Ansell
44337
4X-Large Elastic Wrists, Ankles with Hood and Boots N/A
Only null left
19-120-2401F
View Documents
Ansell
44336
3X-Large Elastic Wrists, Ankles with Hood and Boots N/A
Only null left
19-120-2401C
View Documents
Ansell
44333
Large Elastic Wrists, Ankles with Hood and Boots N/A
Only null left
19-120-2401E
View Documents
Ansell
44335
2X-Large Elastic Wrists, Ankles with Hood and Boots N/A
Only null left
Description
Description
Protective coveralls are made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals, and dry particulates.
  • Breathable, microporous film laminate fabric.
  • Barrier protection against hazardous particulates and aerosols and against low-hazard liquid splash.
  • ASTM F1670 for blood penetration and ASTM F1671 for bloodborne pathogens.
  • ASTM F903 Penetration Testing provides passing results for isocyanate paint (40%) and Potassium Chromate (20%).
  • Anti-stat according to NFPA 99.
  • Features a zippered front and come in a variety of sizes.
  • Eligible for recycling via RightCycle when used in non-hazardous applications as classified by federal, state, and local regulations.
  • Various configurations available, including open or elastic wrists and ankles, elastic back, a hood, and non-skid boots.
  • Human-centric Reflex™ design offers comfortable fit and freedom of movement.
Compliance
Meets ANSI/ISEA 101-1996; ASTM F1670 testing for penetration of blood and bodily fluids; NFPA 99 healthcare facilities criteria for antistatic materials.
Specifications
Specifications
Unisex
Microporous Film Laminate
White
Particle, Liquid
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'