missing translation for 'onlineSavingsMsg'
Learn More

Ansell KleenGuard™ A40 Liquid and Particle Protection Shoe Covers

Catalog No. 19169901
Change view
Click to view available options
Size:
Universal
Large
X-Large
Height (English):
5.25 in.
7 in.
Closure Type:
Elastic Opening
Elastic Top
Fully Elastic
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Height (English) Closure Type
19169901 Universal 7 in. Elastic Opening
191202410 Universal 7 in. Fully Elastic
191202411 Large 5.25 in. Elastic Top
19121719 X-Large 5.25 in. Elastic Top
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19169901 Supplier Ansell Supplier No. 27000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A40 shoe covers are made with breathable, microporous film to protect from debris, non-hazardous liquid chemicals, and dry particulates.

  • Made of microporous film laminate
  • Serged seams
  • Choose from fully elasticized formed fit or elastic opening styles
  • Ambidextrous single use cover 5.25 in. or 7 in. height
  • Made to keep out debris, dry particulates, and some liquids
  • Adheres to NFPA 99 criteria for antistatic materials

Compliance

Meets NFPA™ 99 Health Care Facilities criteria for antistatic materials.

Specifications

Size Universal
Material Microporous Fabric
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Opening
Model 27000
Seam Type Serged
For Use With (Application) Manufacturing, Painting, Cleaning, Liquid Handling, Utilities, Maintenance
Certifications/Compliance NFPA 99
Packaging Quantity 300/Cs.
Height (English) 7 in.
Resistance Fluid
Product Type Shoe Covers
Show More Show Less