Learn More
Ansell KleenGuard™ A40 Liquid and Particle Protection Sleeves
GREENER_CHOICE
Catalog No. 191202406
Catalog No. 191202406 Supplier Ansell Supplier No. 44480
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Breathable, microporous laminate protects against nonhazardous liquids
White, disposable sleeve protectors.
  • 18 in. long (45.7cm) sleeve protectors feature elastic ends for a secure fit
  • Low lint
Compliance
Meet NFPA™ 99 for Antistatic Materials and ASTM™ F1670 testing for penetration of blood and bodily fluids.
Specifications
Size Universal
Material Microporous Film Laminate
For Use With (Application) Fiberglass manufacturing, tank cleaning, utilities, liquid handling, pressure washing, maintenance, general manufacturing
Length (Metric) 45.7 cm