missing translation for 'onlineSavingsMsg'
Learn More

KleenGuard™ A40 Liquid and Particle Protection Aprons
GREENER_CHOICE

Catalog No. 191202405
Change view
Click to view available options
Size:
Universal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
191202405 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 191202405 Supplier KleenGuard™ Supplier No. 44481

KleenGuard™ A40 Liquid and Particle Protection Aprons feature a breathable, microporous film laminate that protects against nonhazardous liquids.

  • White, disposable aprons
  • Ties in the back
  • Breathable, microporous film laminate
  • Excellent liquid and particulate barrier
  • Low in lint

Compliance

Meet NFPA™ 99 for Antistatic Materials and ASTM™ F1670 testing for penetration of blood and bodily fluids.

Specifications

Type Apron
Size Universal
Material 23-P Film Laminate
Color White
Disposable Reusable
Closure Type Ties
Sterility Non-sterile
For Use With (Application) Fiberglass manufacturing, tank cleaning, utilities, liquid handling, pressure washing, maintenance, general manufacturing
Certifications/Compliance NFPA 99, ASTM F1670
Dimensions (L x W) 101.6 x 71 cm (40 x 28 in.)
Length (English) 40 in.
Length (Metric) 101.6 cm
Width (English) 28 in.
Width (Metric) 71 cm
Item Description Kleenguard A40 Light Duty Short Apron - Apron / White
Seam Type Sewn
Show More Show Less