Learn More
Ansell KleenGuard™ A35 Liquid and Particle Protection Coveralls
Provide a comfortable fit while providing liquid and particulate protection
Supplier:  Ansell 38932
 View more versions of this product
Catalog No. 19-006-216
Only null left
Explore available promotions
Description
Description
  • Breathable
  • Microporous film laminate
  • Lightweight outer fabric that protects the microporous film laminate
  • Added durability and comfort liquid barrier
  • Particulate barrier
  • Available with elastic wrists and ankles
  • With or without hood
  • Zipper front
Specifications
Specifications
General Purpose Coveralls
Unisex
White
Elastic
Elastic
Unhooded
Serged
Dust, Liquid
KleenGuard™ A35
Elastic Wrists and Ankles
3X-Large
Microporous Film Laminate
Limited-use
Elastic
Collared
Front Zipper
Non-sterile
ANSI/ISEA 101-2014
General Industrial, Manufacturing, Laboratory Work, Painting, Maintenance
Product Suggestions
Product Suggestions