missing translation for 'onlineSavingsMsg'
Learn More

Ansell KleenGuard™ A35 Liquid and Particle Protection Coveralls
GREENER_CHOICE

Catalog No. 17986026
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Style:
Elastic Wrists and Ankles
Elastic Wrists and Ankles with Attached Hood and Boots
Elastic Wrists, Ankles, and Hood
Open Wrists and Ankles, No Elastic
31 product options available for selection
Product selection table with 31 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Style
17986026 Medium Elastic Wrists and Ankles with Attached Hood and Boots
17986029 2X-Large Elastic Wrists and Ankles with Attached Hood and Boots
17986025 Small Elastic Wrists and Ankles with Attached Hood and Boots
17986027 Large Elastic Wrists and Ankles with Attached Hood and Boots
17986031 4X-Large Elastic Wrists and Ankles with Attached Hood and Boots
17986030 3X-Large Elastic Wrists and Ankles with Attached Hood and Boots
17986028 X-Large Elastic Wrists and Ankles with Attached Hood and Boots
19006211 Small Elastic Wrists and Ankles
19006212 Medium Elastic Wrists and Ankles
19006213 Large Elastic Wrists and Ankles
19006214 X-Large Elastic Wrists and Ankles
19006215 2X-Large Elastic Wrists and Ankles
19006216 3X-Large Elastic Wrists and Ankles
19006217 4X-Large Elastic Wrists and Ankles
19006220 Small Open Wrists and Ankles, No Elastic
296364457 5X-Large Elastic Wrists and Ankles
19006221 Medium Open Wrists and Ankles, No Elastic
19006222 Large Open Wrists and Ankles, No Elastic
19006223 X-Large Open Wrists and Ankles, No Elastic
19006224 2X-Large Open Wrists and Ankles, No Elastic
19006225 3X-Large Open Wrists and Ankles, No Elastic
19006226 4X-Large Open Wrists and Ankles, No Elastic
19006227 5X-Large Open Wrists and Ankles, No Elastic
19162580 Small Elastic Wrists, Ankles, and Hood
19162581 Medium Elastic Wrists, Ankles, and Hood
19162582 Large Elastic Wrists, Ankles, and Hood
19162583 X-Large Elastic Wrists, Ankles, and Hood
19162584 2X-Large Elastic Wrists, Ankles, and Hood
19162586 4X-Large Elastic Wrists, Ankles, and Hood
19162587 5X-Large Elastic Wrists, Ankles, and Hood
19162585 3X-Large Elastic Wrists, Ankles, and Hood
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 31 options available.
31 options
Catalog No. 17986026 Supplier Ansell Supplier No. 38948

Provide a comfortable fit while providing liquid and particulate protection

  • Breathable
  • Microporous film laminate
  • Lightweight outer fabric that protects the microporous film laminate
  • Added durability and comfort liquid barrier
  • Particulate barrier
  • Available with elastic wrists and ankles
  • With or without hood
  • Zipper front

Specifications

Product Type General Purpose Coveralls
Size Medium
Gender Unisex
Material Microporous Film Laminate
Color White
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic, Attached Boots
Cuff Style Elastic
Neck Style Hooded
Hood Type Elastic
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Dust, Liquid
Certifications/Compliance ANSI/ISEA 101-2014
Model KleenGuard™ A35
For Use With (Application) General Industrial, Manufacturing, Laboratory Work, Painting, Maintenance
Style Elastic Wrists and Ankles with Attached Hood and Boots
Show More Show Less