missing translation for 'onlineSavingsMsg'
Learn More

Ansell KleenGuard™ A20 Breathable Particle Protection Lab Coats
GREENER_CHOICE

Catalog No. 19457323
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Closure Type:
Hook and Loop
Snaps
No. of Pockets:
2
None
Cuff Style:
Elastic
Open
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Closure Type No. of Pockets Cuff Style
19457323 Large Hook and Loop None Elastic
19457321 Small Hook and Loop None Elastic
19457322 Medium Hook and Loop None Elastic
19457324 X-Large Hook and Loop None Elastic
19457325 2X-Large Hook and Loop None Elastic
19457326 3X-Large Hook and Loop None Elastic
19457327 4X-Large Hook and Loop None Elastic
19457328 5X-Large Hook and Loop None Elastic
01349B Medium Snaps 2 Open
01349C Large Snaps 2 Open
01349D X-Large Snaps 2 Open
296355656 2X-Large Snaps 2 Open
17010302 3X-Large Snaps 2 Open
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 19457323 Supplier Ansell Supplier No. 35620
Edge

A20 lab coats are comprised of five layers of stacked barrier protection (SMMMS) against hazardous particles. They are a popular choice for dusty environments such as laboratories and remediation, as well as pesticide applications and as decontamination modesty garments.

  • Tailored style made of breathable patented MICROFORCE™ Barrier SMS Fabric
  • Knee-length, serged seams
  • Long sleeves, notched-lapel collar
  • Air and moisture vapor can pass through to aid wearer comfort
  • Meets NFPA 99 criteria for antistatic materials
  • Two styles to choose from:
    • Open wrists with pockets, snap closure
    • Elastic wrists without pockets, hook and loop closure

Compliance

NFPA 99 compliant antistatic material

Specifications

Product Type General Purpose Lab Coat
Size Large
Gender Unisex
Material SMS
Color White
Closure Type Hook and Loop
Cuff Style Elastic
Disposable Limited-use
Sterility Non-sterile
Style Knee-length
No. of Pockets None
Collar Type Notched Lapel
Model 35620
Seam Type Serged
Certifications/Compliance NFPA 99
For Use With (Application) Laboratories, Remediation, Decontamination, Pesticides
Construction 1.6 oz. Fabric
Show More Show Less