missing translation for 'onlineSavingsMsg'
Learn More

Ansell KleenGuard™ A20 Blue Coveralls, Attached Hood and Boots

Catalog No. 29633849
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3X-Large
4X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
29633849 Large
29633850 X-Large
29633851 2X-Large
29633852 3X-Large
296352222 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 29633849 Supplier Ansell Supplier No. 58523

Denim blue color coveralls feature a breathable and abrasion-resistant fabric to keep workers cool and comfortable while protecting against fibers, fine dusts, and low surface tension liquid splashes.

  • Patented Reflex™ design is 7.5 times less likely to rip out than ANSI minimums.
  • Provides 12% more chest room and 6% longer body length.
  • Ideal for use in dusty environments, as well as for painting, manufacture spraying and agricultural spraying.
  • Stylish denim blue color masks dirt and grease.
  • Air and moisture vapor can pass through to aid wearer comfort.
  • Triple-stitched serged seams.
  • Can withstand multiple wearing.
  • Zippered front, elastic back, attached hood and boots.

Specifications

Product Type General Purpose Coveralls
Size Large
Gender Unisex
Material SMS Fabric
Color Denim Blue
Disposable Limited-use
Ankle Style Elastic, Attached Boots
Cuff Style Elastic
Neck Style Hooded
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Dust
Back Style Elastic
Certifications/Compliance NFPA 99
Description Microforce™ Barrier SMS Fabric
Model KleenGuard™ A20
For Use With (Application) Laboratories, Decontamination Modesty Garment, Remediation, Pesticides, Heavy Dust Environments
Show More Show Less