Learn More
Ansell KleenGuard™ A20 Coveralls, Elastic Back, Wrists and Ankles with Attached Hood
Coveralls are designed to protect against particles such as dirt, grime, and dust. Made with breathable SMS fabric with five layers of stacked barrier protection.
Supplier:  Ansell 49113
 View more versions of this product
Catalog No. 17-981-47A
Only null left
Explore available promotions
Alert:
Not for use near heat, flames or flammable environments
Description
Description
  • For use in laboratories, remediation, decontamination, pesticides, and heavy dust environments.
  • Elastic back, wrists and ankles with attached hood, zippered front.
  • Provides general protection against many particles
  • Breathable patented Microforce™ barrier SMS fabric passes NFPA 99 for antistatic materials.
  • High abrasion resistance, low linting
  • Withstands multiple wearings
  • Air and moisture vapor can pass through to aid wearer comfort.
  • Triple-stitched serged seams.
  • Offer particle holdout coupled with unmatched comfort attributes.
  • Fabric is comprised of five layers of stacked barrier protection (SMMMS).
  • Human-centric Reflex™ design offers comfortable fit and freedom of movement.
  • Eligible for recycling via RightCycle™ when used in non-hazardous applications as classified by federal, state, and local regulations.
Specifications
Specifications
General Purpose Coveralls
Unisex
White
Elastic
Hooded
Non-sterile
Elastic
Microforce™ Barrier SMS Fabric
Laboratories, Decontamination Modesty Garment, Remediation, Pesticides, Heavy Dust Environments
Large
SMS Fabric
Limited-use
Elastic
Front Zipper
Dust
NFPA 99
KleenGuard™ A20
Product Suggestions
Product Suggestions