missing translation for 'onlineSavingsMsg'
Learn More

Ansell KleenGuard™ A20 Coveralls, Elastic Back, Wrists and Ankles with Attached Hood
GREENER_CHOICE

Catalog No. 1798147E
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
1798147E Medium
2870728 2X-Large
19812952 4X-Large
1798147A Large
1798147B X-Large
1798147D 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Not for use near heat, flames or flammable environments
Catalog No. 1798147E Supplier Ansell Supplier No. 49112

missing translation for 'validMainframeMsg'

Not for use near heat, flames or flammable environments

Coveralls are designed to protect against particles such as dirt, grime, and dust. Made with breathable SMS fabric with five layers of stacked barrier protection.

  • For use in laboratories, remediation, decontamination, pesticides, and heavy dust environments.
  • Elastic back, wrists and ankles with attached hood, zippered front.
  • Provides general protection against many particles
  • Breathable patented Microforce™ barrier SMS fabric passes NFPA 99 for antistatic materials.
  • High abrasion resistance, low linting
  • Withstands multiple wearings
  • Air and moisture vapor can pass through to aid wearer comfort.
  • Triple-stitched serged seams.
  • Offer particle holdout coupled with unmatched comfort attributes.
  • Fabric is comprised of five layers of stacked barrier protection (SMMMS).
  • Human-centric Reflex™ design offers comfortable fit and freedom of movement.
  • Eligible for recycling via RightCycle™ when used in non-hazardous applications as classified by federal, state, and local regulations.

Specifications

Product Type General Purpose Coveralls
Size Medium
Gender Unisex
Material SMS Fabric
Color White
Disposable Limited-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Front Zipper
Sterility Non-sterile
Resistance Dust
Back Style Elastic
Certifications/Compliance NFPA 99
Description Microforce™ Barrier SMS Fabric
Model KleenGuard™ A20
For Use With (Application) Laboratories, Decontamination Modesty Garment, Remediation, Pesticides, Heavy Dust Environments
Show More Show Less