missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell KleenGuard™ A20 Breathable Particle Protection Coveralls, Elastic Back, Wrists, and Ankles

Description
- For use in laboratories, remediation, decontamination, pesticides, and heavy dust environments.
- Elastic back, wrists and ankles, zippered front.
- Provides general protection against many particles
- Breathable patented Microforce™ barrier SMS fabric passes NFPA 99 for antistatic materials.
- High abrasion resistance, low linting
- Withstands multiple wearings
- Air and moisture vapor can pass through to aid wearer comfort.
- Triple-stitched serged seams.
- Offer particle holdout coupled with unmatched comfort attributes.
- Fabric is comprised of five layers of stacked barrier protection (SMMMS).
- Human-centric Reflex™ design offers comfortable fit and freedom of movement.
- Eligible for recycling via RightCycle™ when used in non-hazardous applications as classified by federal, state, and local regulations.
Specifications
Specifications
| Product Type | General Purpose Coveralls |
| Size | X-Large |
| Gender | Unisex |
| Material | SMS Fabric |
| Color | White |
| Disposable | Limited-use |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Closure Type | Front Zipper |
| Sterility | Non-sterile |
| Show More |