missing translation for 'onlineSavingsMsg'
Learn More

Ansell KleenGuard™ A20 Boot Covers
GREENER_CHOICE

Catalog No. 013496
Change view
Click to view available options
Closure Type:
Elastic Top
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Type
013496 Elastic Top
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 013496 Supplier Ansell Supplier No. 36880
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Manufactured to the highest standards. KleenGuard™ A20 Boot Covers pass NFPA 99 criteria for antistatic materials.

  • Made of white, breathable patented Microforce™ Barrier SMS Fabric
  • Covers fit over virtually all boots
  • With elastic tops

Boot covers may not provide traction on slippery floors.

Specifications

Size Universal
Material SMS
Color White
Closure Type Elastic Top
For Use With (Application) Laboratories, Decontamination, Remediation, Pesticides, Heavy Dust Environments
Certifications/Compliance NFPA 99
Product Type Shoe Covers