missing translation for 'onlineSavingsMsg'
Learn More

Kimtech™ A8 Non-Sterile Cleanroom Shoe Covers, Unitrax
GREENER_CHOICE

Catalog No. 19169651
Change view
Click to view available options
Size:
Small/Medium
Universal
X-Large/2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19169651 Universal
19169652 X-Large/2X-Large
19169650 Small/Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19169651 Supplier Kimtech™ Supplier No. 39371

Seamless, non-skid Kimtech™ A8 Non-Sterile Cleanroom Shoe Covers are low-lint for contamination control in environments where wet splash and dry particle protection are needed.

  • 300 Kimtech™ A8 Shoe Covers / Case, 3 Bags of 100 Covers; White
  • Superior non-skid design has 4 times more grip than leading alternatives
  • Seamless bottom for better comfort; elastic ankle opening
  • Low lint for contamination control
  • Not made with natural rubber latex
  • Made with poly-film laminated fabric
  • Double bagged for use in clean manufacturing and quality control environments
  • Elastic ankle opening for a snug fit
  • Recyclable through the RightCycle Program, which turns used lab coats into raw materials for eco-friendly consumer products

Specifications

Size Universal
Material Polyethylene, Polyester
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Non-skid
Seam Type Seamless
For Use With (Application) Cleanrooms, Manufacturing
Packaging Quantity 300/Cs.
Packaging Type Double-bagged, 3 Bags of 100 Covers
Resistance None
Product Type Controlled Environments Shoe Covers
Show More Show Less