Learn More
Ansell Kimtech™ A7 Cleanroom Lab Coats
GREENER_CHOICE
Catalog No. 19169106
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
4X-Large
6X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19169106 Small
19169107 Medium
19169108 Large
19169109 X-Large
19169110 2X-Large
19169111 4X-Large
19169112 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19169106 Supplier Ansell Supplier No. 47651
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Lab coats provide protection and prevent contamination from liquid splashes in controlled environments.
  • Available in bulk cleanroom packaging (double bag)
  • Anti-static treatment and are low lint to reduce contamination risk
  • High collar for increased neck protection
  • Comfortable thumb loops to maintain glove and garment interface
  • Disposable lab coats are recyclable through the RightCycle Program
  • Enhanced chemical and biological protection (passes ASTM F1670 for penetration of blood and body fluids)
  • Protection for people and processes against liquid splashes
Specifications
Product Type Controlled Environments Lab Coat
Size Small
Material Polypropylene
Color White
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
Resistance Static
Collar Type High Mandarin Collar
Latex Free Yes
Seam Type Serged, Triple Overlock Stitching
Certifications/Compliance ASTM F1670
Certifications/Compliance (Europe) EN 1149-5:2018 Type 6 Limited Splash, EN 1149-1:1995 - Antistatic; CE 0120
Includes Thumb Loop
Packaging Type Bulk, Double Bagged
Show More Show Less