Learn More
Kimtech™ A5 Sterile Hood
GREENER_CHOICE
Catalog No. 19169867
Change view
Click to view available options
Closure Type:
None
Ties
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Type
19169867 Ties
19057495 None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19169867 Supplier Kimtech™ Supplier No. 25797
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Sterile cleanroom hoods feature innovative CLEAN-DON™ technology for simpler aseptic donning. Kimtech™ A5 Sterile Hood suitable for EU GMP ISO 5 Grade A sterile cleanrooms and individually vacuum-packed.
  • Suitable for use in EU GMP ISO 5 Class A sterile cleanroom environments and laboratories
  • Advanced CLEAN-DON™ technology for fast, simple and compliant aseptic donning that combines two donning steps into one
  • Comfortable hoods are provided in a universal size
  • Made from low-lint breathable SMS fabric with a cloth-like feel
  • Contamination protection is ensured through high quality sterility assured material with high-strength bound and triple-stitched seams, and secure ties
  • Individually vacuum-packed and efficiently double-bagged to save space
  • PPE Cat III according to Regulation (EU) 2016/425
  • Type 6 (PB) certified
  • Certificate of Conformance by lot available online
  • Certificate of Sterility by lot available online
Order Info
Gamma sterilized and individually bagged. Certificate of Irradiation available.
Specifications
Product Type Open Face
Size Universal
Material Polypropylene
Color White
Closure Type Ties
Disposable Single-use
Latex Free Yes
Model 25797
Sterility Sterile
Packaging Type Triple-bagged, Vacuum Packaged
Quantity 100/Cs.
Includes With Ties
Cleanroom Class Class 100 (ISO 5)
Show More Show Less