Learn More
Kimtech™ A5 Sterile Cleanroom Boots, Grasp Ties and Vinyl Edge
GREENER_CHOICE
Catalog No. 19169902
Change view
Click to view available options
Size:
Small/Medium
Universal
X-Large/2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19169902 Small/Medium
19169903 Universal
19169904 X-Large/2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19169902 Supplier Kimtech™ Supplier No. 31683
Sterile cleanroom boots with a wrap-around vinyl foot feature innovative CLEAN-DON™ technology for aseptic donning. Kimtech™ A5 Sterile Cleanroom Boots with Grasp Ties and Vinyl Edge suitable for EU GMP ISO 5 Grade A sterile cleanrooms.
  • Sterile cleanroom boots with grasp ties and vinyl edge
  • Average bacterial filtration efficiency (BFE) of 96%
  • Made of 100% polypropylene breathable SMS fabric
  • Triple-bagged and vacuum packaged
  • Suitable for ISO Class 5 or higher cleanrooms
  • Clean-Don ties and 2-inch edge vinyl, for an enhance moisture barrier
  • Made without latex, silicone or the BHT preservative and are designed for single-use
  • 100 Pairs/200 Each
  • CE certified (CE Simple)
  • Each individually bagged pair of boot covers has a sterilization indicator as a visual that it's undergone gamma sterilization
  • A5 Cleanroom Apparel is manufactured in an ISO-certified facility, clean-processes and individually packaged under cleanroom conditions
Specifications
Size Small/Medium
Material SMS
Latex Free Yes
Color White
Disposable Single-use
Sterility Sterile
Closure Type Side-grasp Ties
Sole Type Non-skid
Packaging Quantity 100 Pairs/Cs.
Packaging Type Triple Bagged, Vacuum Packed
Resistance None
Product Type Controlled Environments Boot Covers
Cleanroom Class Class 100 (ISO 5)
Show More Show Less