missing translation for 'onlineSavingsMsg'
Learn More

Ansell Kimtech Pure™ A5 Cleanroom Non-Sterile Hoods
GREENER_CHOICE

Catalog No. 19169868
Change view
Click to view available options
Closure Type:
None
Ties
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Type
19169868 Ties
19812863 None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19169868 Supplier Ansell Supplier No. 25796

Non-sterile cleanroom hoods cover the head, neck, and shoulders to prevent contamination in the environment. Availble with or without ties.

  • Non-sterile, white
  • Triple bagged, vacuum packed
  • Suitable for ISO Class 5 or higher cleanrooms; CE certified
  • Works with a variety of masks
  • Latex-free
  • Kimtech™ A5 Cleanroom Apparel is manufactured in an ISO-certified facility, clean-processes and individually packaged under cleanroom conditions

Specifications

Product Type Open Face
Size Universal
Material Polypropylene SMS Fabric
Color White
Closure Type Ties
Latex Free Yes
Model 25796
Sterility Non-sterile
Packaging Type Triple-bagged, Vacuum Packaged
Quantity 100/Cs.
Includes With Ties
For Use With (Application) ISO 5 to 8 cleanroom environments, compounding, mixing, filling, cleaning
Cleanroom Class Class 100 (ISO 5)
Show More Show Less