missing translation for 'onlineSavingsMsg'
Learn More

Ansell Kimtech™ A5 Cleanroom Apparel, Bulk Packaging
GREENER_CHOICE

Catalog No. 19169052
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large, 7X-Large, 8X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19169052 Small
19169053 Medium
19169054 Large
19169055 X-Large
19169056 2X-Large
19169057 3X-Large
19169058 4X-Large
19169059 5X-Large
19169060 6X-Large, 7X-Large, 8X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19169052 Supplier Ansell Supplier No. 49831

Coveralls feature a polypropylene breathable SMS fabric with a cloth-like feel.

Kimtech™ A5 Cleanroom Coveralls are made of 100% polypropylene breathable SMS fabric, which gives the wearer cloth-like comfort. While other coveralls for cleanroom environments may look similar, the ones from Kimtech™ Pure have distinct advantages including tightly bound seams to strengthen the barrier, a high neck collar to reduce contamination, thumb loops to keep the sleeves from rolling back and a zipper flap to cover the zipper area.

In addition, this protective apparel is recyclable through the RightCycle program.

This personal protective apparel is triple-bagged and vacuum packaged, suitable for ISO Class 5 or higher cleanrooms. It is not made with natural rubber latex, silicone, or the BHT preservative and is designed for single-use (disposable PPE).

For use in biotech, medical device, electronics and telecommunications manufacturing, they are designed to protect your process by providing comfortable, sustainable protection.

  • Made of breathable SMS fabric, the coveralls feature a high-neck collar, a zipper flap, thumb loops and bound seams
  • Triple bagged, vacuum packed, suitable for ISO Class 5 or higher cleanrooms; CE certified
  • Kimtech™ A5 Cleanroom Apparel is manufactured in an ISO-certified facility, clean-processes and individually packaged under cleanroom conditions
  • Recyclable through the RightCycle program, which can help you meet your sustainability goals

Specifications

Product Type Cleanroom Coveralls
Size Small
Material Polypropylene, SMS
Color White
Disposable Single-use
Cuff Style Thumb Loop
Neck Style Collared
Sterility Non-sterile
Collar Type Standing
For Use With (Application) Compounding, Mixing, Filling, Cleaning, Pharmaceutical, Biomedial Research
Cleanroom Class Class 100-100,000 (ISO 5-8)
Show More Show Less