missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimberly-Clark Professional™ Scott™ Control Antimicrobial Foam Skin Cleanser (0.1% Benzalkonium Chloride)

Contains (0.1% Benzalkonium Chloride) to kill harmful germs such as E. Coli, Listeria, and Salmonella. Contains aloe and glycerin to moisturize the skin. Does not contain triclosan. High capacity for less refilling and maintenance time. Easy-to-load cassette clicks right into place. Bottle is recyclable after the pump is removed.

$94.18 - $94.18

Specifications

Color Clear
Quantity 6 Pk.
Volume (English) 0.26 gal.
Volume (Metric) 1 L
View More Specs

Products 1
Catalog Number Mfr. No. Product Type Volume (English) Quantity Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Product Type Volume (English) Quantity Volume (Metric) Price Quantity  
19-413-007
View Documents Promotion Details
Kimberly-Clark Professional™
91554
Anti-bacterial Foam Skin Cleanser 0.26 gal. 6 Pk. 1 L
Case of 6 Each for $94.18
 
Description

Description

  • 6 cassettes / case; 1.0 L each; Scott Control E-2 Antimicrobial Foam Skin Cleanser contains 0.1% Benzalkonium Chloride and kills 99.9% of the most germs in as little as 15 seconds
  • It kills germs including Salmonella, Staphylococcus aureaus, and E. coli
  • This foaming commercial hand soap clear (dye-free), has a pleasant fruit scent
  • Easy-to-load, high capacity cassette clicks right into place and is compatible with the Scott Control Professional Cassette Skin Care Dispenser
  • The bottle is recyclable after the pump is removed

Specifications

Specifications

Clear
6 Pk.
0.26 gal.
1 L
SDS
missing translation for 'documents'

missing translation for 'documents'