Learn More
Ansell KleenGuard™ V90 Shield™ Safety Goggles with Face Shield
Catalog No. 296362115
Change view
Click to view available options
Lens Tint:
Clear
Catalog No. Lens Tint
296362115 Clear
1 options
Catalog No. 296362115 Supplier Ansell Supplier No. 18629
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
When you have a job to tackle that involves flying particles or splashes, wearing safety glasses isn't enough. Protect yourself with the KleenGuard V90 Shield Safety Goggles with Face Shield.
  • MonoGoggle XTR with full-face protection
  • Shield detaches from goggle
  • Blue frame with Visiclear Clear lens
  • Replacement shield available (see cat. no. 172831)
Compliance
Meets ANSI Z87.1+ 2010 standards.
Specifications
Certifications/Compliance ANSI Z87.1-2010
Lens Tint Clear
Product Type Safety Goggles
For Use With (Application) Provides full eye and face protection from liquids without primary eye protection underneath a face shield
Frame Color Blue
Material Polycarbonate
UV Protection UV400