Learn More
Ansell KleenGuard™ V10 Element™ Eyewear
GREENER_CHOICE
Catalog No. 19706002
Change view
Click to view available options
Frame Color:
Clear
Lens Tint:
Clear
Catalog No. Frame Color Lens Tint
19706002 Clear Clear
1 options
Catalog No. 19706002 Supplier Ansell Supplier No. 25627
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
When you need an economical way to provide on-the-job eye protection, KleenGuard Element Safety Eyewear is an excellent choice.
  • Lightweight design
  • Universal nose bridge for comfort
  • Base curve 9.5
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
Certifications/Compliance Meets ANSI Z87.1-2010 standards.
Lens Tint Clear
Frame Color Clear
No. per Pack 12/Pk.