Learn More
KleenGuard™ G60 Purple Nitrile™ Cut-Resistant Gloves
Cut-resistant gloves are constructed of comfortable, breathable fabric and feature purple tapered coating on the fingertips for added dexterity.
Supplier:  KleenGuard™ 97431
 View more versions of this product
Catalog No. 19-161-593
Only null left
Explore available promotions
Description
Description
High level of cut resistance while enhancing dexterity, grip and comfort.
  • Dyneema™ fabric construction
  • Purple Nitrile coated fingertips
  • Flexible dotted palm
  • Ambidextrous
Specifications
Specifications
Nitrile Dots
ANSI A3
Yes
Cut-resistant Gloves
EN 388
Knit
Kleenguard™ G60
White, Purple
Ambidextrous
Dyneema
8
Glass/Metal handling, Paper making and automotive/aircraft assembly
97431
Product Suggestions
Product Suggestions