missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ Access™ Wiper Dispenser

Catalog No. 17228C
Change view
Click to view available options
Mounting:
Wall or Surface Mount
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
17228C Wall or Surface Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17228C Supplier Kimberly-Clark Professional™ Supplier No. 73900

Space-saving metal dispenser

Mounts to walls, workbenches, trucks, carts and holds one box of:

  • WypAll™ WypAll X80 towels: 19-130-1631, 19-130-165, 19-130-2379 or 19-130-3474
  • WypAll L20 Wipers: 06-666-19 and 06-666-22
  • Kimtech Science Lens Cleaning Stations: 17-228A
  • WypAll L40 multi-purpose wipers: 06-666-130 L30
  • WypAll L40 general-purpose wipers: 19-168-202
  • WypAll X70 wipers: 19-014-872, 19-812-925 and 19-014-860

Specifications

Product Type Wiper Dispenser
Color White
Dimensions (L x W x H) 11 x 0.65 x 8 in.
Mounting Wall or Surface Mount
For Use With (Equipment) For WYPALL wiper boxes