missing translation for 'onlineSavingsMsg'
Learn More

Ansell KleenGuard™ A20 Frocks
GREENER_CHOICE

Catalog No. 19457221
Change view
Click to view available options
Size:
5X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19457221 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19457221 Supplier Ansell Supplier No. 34815

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Frocks feature breathable, patented MICROFORCE barrier SMS fabric and meets NFPA 99 criteria for antistatic materials.

  • Snap front closure with snap at collar
  • Long length
  • Elastic wrists
  • No pockets
  • Passes NFPA 99 criteria for antistatic materials
  • Typical uses: Laboratories; Decontamination modesty garment; Remediation; Pesticides; Heavy dust environments

Order Info

This size is available on a made-to-order basis – 30 case minimum order per product code; product available within 8 weeks of order receipt

Compliance

Passes NFPA 99 criteria for antistatic materials

Specifications

Size 5X-Large
Material SMS
Color White
Closure Type Snap (Front)
Cuff Style Elastic
Certifications/Compliance NFPA 99
Product Type General Purpose Frock
For Use With (Application) Laboratories, Decontamination, Remediation, Pesticides, Heavy Dust Environments