missing translation for 'onlineSavingsMsg'
Learn More

Ansell KleenGuard™ A20 Breathable Particle Protection Frocks
GREENER_CHOICE

Catalog No. p-24498
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
191501613 Medium
191501615 2X-Large
191501614 Large
19161734 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 191501613 Supplier Ansell Supplier No. 36262
Only null left
Edge

Protection against dry particulates and light liquid sprays

  • Long, abrasion-resistant jackets made of breathable SMMMS fabric - 5 layers of stacked barrier protection
  • Raglan sleeves with elastic wrists and high collar
  • No pockets
  • Plastic snap front closure
  • White

Specifications

Size Medium
Material SMS
Color White
Closure Type Snaps
For Use With (Application) Suited for laboratories, decontamination modesty garment, remediation, pesticides, heavy dust environments
Certifications/Compliance NFPA 99
Length (English) 40 in.
Length (Metric) 102 cm