missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell KleenGuard™ A20 Blue Coveralls with Elastic Back, Wrists, Ankles, and Hood
Denim blue color coveralls are designed to protect against particles such as dirt, grime, and dust. Made with breathable SMS fabric with five layers of stacked barrier protection.
Supplier: Ansell 58515
Description
- For use in laboratories, remediation, decontamination, pesticides, and heavy dust environments.
- Elastic back, wrists and ankles with attached hood, zippered front.
- Provides general protection against many particles
- Breathable patented Microforce™ barrier SMS fabric passes NFPA 99 for antistatic materials.
- High abrasion resistance, low linting
- Withstands multiple wearings
- Air and moisture vapor can pass through to aid wearer comfort.
- Triple-stitched serged seams.
- Offer particle holdout coupled with unmatched comfort attributes.
- Fabric is comprised of five layers of stacked barrier protection (SMMMS).
- Human-centric Reflex™ design offers comfortable fit and freedom of movement.
- Eligible for recycling via RightCycle™ when used in non-hazardous applications as classified by federal, state, and local regulations.
Specifications
| General Purpose Coveralls | |
| Unisex | |
| Denim Blue | |
| Elastic | |
| Hooded | |
| Front Zipper | |
| Non-sterile | |
| Elastic | |
| Microforce™ Barrier SMS Fabric | |
| Laboratories, Decontamination Modesty Garment, Remediation, Pesticides, Heavy Dust Environments |
| 2X-Large | |
| SMS Fabric | |
| Limited-use | |
| Elastic | |
| Attached | |
| Serged | |
| Dust | |
| NFPA 99 | |
| KleenGuard™ A20 |