missing translation for 'onlineSavingsMsg'
Learn More

Ansell KleenGuard™ A20 Blue Coveralls with Elastic Back, Wrists, Ankles, and Hood

Catalog No. 29633844
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
29633844 Large
29633846 2X-Large
29633845 X-Large
29633843 Medium
190881931G 4X-Large
29633847 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 29633844 Supplier Ansell Supplier No. 58513

missing translation for 'validMainframeMsg'

Denim blue color coveralls are designed to protect against particles such as dirt, grime, and dust. Made with breathable SMS fabric with five layers of stacked barrier protection.

  • For use in laboratories, remediation, decontamination, pesticides, and heavy dust environments.
  • Elastic back, wrists and ankles with attached hood, zippered front.
  • Provides general protection against many particles
  • Breathable patented Microforce™ barrier SMS fabric passes NFPA 99 for antistatic materials.
  • High abrasion resistance, low linting
  • Withstands multiple wearings
  • Air and moisture vapor can pass through to aid wearer comfort.
  • Triple-stitched serged seams.
  • Offer particle holdout coupled with unmatched comfort attributes.
  • Fabric is comprised of five layers of stacked barrier protection (SMMMS).
  • Human-centric Reflex™ design offers comfortable fit and freedom of movement.
  • Eligible for recycling via RightCycle™ when used in non-hazardous applications as classified by federal, state, and local regulations.

Specifications

Product Type General Purpose Coveralls
Size Large
Gender Unisex
Material SMS Fabric
Color Denim Blue
Disposable Limited-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Dust
Back Style Elastic
Certifications/Compliance NFPA 99
Description Microforce™ Barrier SMS Fabric
Model KleenGuard™ A20
For Use With (Application) Laboratories, Decontamination Modesty Garment, Remediation, Pesticides, Heavy Dust Environments
Show More Show Less