missing translation for 'onlineSavingsMsg'
Learn More

Ansell KleenGuard™ A10 Light Duty Shoe Covers
GREENER_CHOICE

Catalog No. p-3202983
Change view
Click to view available options
Size:
Universal
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19057428 Universal
19999980 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19057428 Supplier Ansell Supplier No. 36811
Only null left

For a wide range of work environments such as food processing, manufacturing facilities, hospitals, and construction sites.

  • Made of lightweight spunbond polypropylene material
  • Fully elasticized and keep workers cool and comfortable
  • Blue
  • Universal size

Specifications

Size Universal
Material Polypropylene
Coating Polyethylene
Color Light Blue
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Low-skid
Model 36811
Seam Type Serged
For Use With (Application) Food Processing, Manufacturing Facilities, Maintenance, Healthcare
Certifications/Compliance ANSI/ISEA 101-1996
Packaging Quantity 300/Cs.
Product Type Shoe Covers
Show More Show Less