missing translation for 'onlineSavingsMsg'
Learn More

Ansell KleenGuard™ A10 Light Duty Beard Cover
GREENER_CHOICE

Catalog No. 19062281
Change view
Click to view available options
Size:
Universal
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19062281 X-Large
19047053 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19062281 Supplier Ansell Supplier No. 66816

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal for food processing, general maintenance, and manufacturing

  • Comfortable
  • Lightweight polypropylene material

Specifications

Size X-Large
Material Polypropylene
Color White
Disposable Single-use
For Use With (Application) Food Processing, General Maintenance, and Manufacturing
Product Type General Purpose Beard Cover