missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell KleenGuard™ A10 Light Duty Beard Cover

Description
- Comfortable
- Lightweight polypropylene material
Specifications
Specifications
| Size | Universal |
| Material | Polypropylene |
| Color | White |
| Disposable | Single-use |
| For Use With (Application) | Food Processing, General Maintenance, and Manufacturing |
| Product Type | General Purpose Beard Cover |