missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell KleenGuard™ A10 Coveralls
A10 Coveralls provide economical, disposable protection for workers exposed to dirt and grime in the workplace.
$138.51 - $138.51
Specifications
| Product Type | General Purpose Coveralls |
|---|---|
| Gender | Unisex |
| Material | Polypropylene |
| Color | White |
| Disposable | Single-use |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
19-041-845
|
Ansell
12230 |
4X-Large |
Case of 25 Each for $138.51
|
|
|||||
Description
- Designed for entry-level protection from hazardous dust
- Feature a zipper front, attached hood, serged seams and elastic wrists
- Five layers of stacked barrier protection, for a lightweight, breathable protective coverall
- Ideal for food processing, general maintenance and manufacturing
Specifications
| General Purpose Coveralls | |
| Polypropylene | |
| Single-use | |
| Elastic | |
| Front Zipper | |
| Non-sterile | |
| Yes |
| Unisex | |
| White | |
| Elastic | |
| Hooded | |
| Serged | |
| ANSI ISEA 101-1996 | |
| Food Processing, General Maintenance, Manufacturing |