missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ Kimtech Science™ Kimwipes™ Delicate Task Wipers, 1-Ply

Easily wipe up liquid and dust. Kimtech Science™ Kimwipes™ Delicate Task Wipers can handle a variety of delicate tasks. Their anti-static dispensing reduces lint and electrostatic discharge and controls usage.

Supplier:  Kimberly-Clark Professional™ 34155

 View more versions of this product

Catalog No. 06-666A



Only null left
Explore available promotions

Description

Description

Easily wipe up liquid and dust. Kimtech Science™ Kimwipes™ Delicate Task Wipers can handle a variety of delicate tasks. Their anti-static dispensing reduces lint and electrostatic discharge and controls usage.

  • Single ply, light-duty wiper that can handle a variety of delicate tasks
  • Extra low-lint wipers made from 100% virgin wood fibers
  • Soft, nonabrasive cellulose-fiber wipers won't scratch most delicate surfaces
  • Easily wipe up liquid and dust
  • Absorb many times their own weight
  • Handy one-at-a-time dispensing packs with special plastic guard to reduce airborne lint, keep contaminants out of box and reduce electrostatic discharge
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications

Specifications

Delicate Task Wipe
20.83 cm
11.17 cm
White
8.2 x 4.4 in.
286 Wipes/Box, 60 Boxes/Cs.
8.2 in.
4.4 in.
Cellulose
Single-use
General-Purpose
1-ply
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions