Learn More
Kimtech™ M7 Face Veils
GREENER_CHOICE
Catalog No. 191600417
Change view
Click to view available options
Fastening Type:
Headband
Snaps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fastening Type
191600417 Headband
19165385 Snaps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 191600417 Supplier Kimtech™ Supplier No. 62757
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Disposable face veils with knitted headbands or snaps are constructed of low-lint polyethylene film. Covered nose piece provides increased comfort. Suitable for ISO Class 7 or higher cleanroom environments.
  • Constructed of low-lint polyethylene film
  • Covered nose piece to provide increased comfort
  • Ultra sonic bond for higher product integrity
  • Latex and silicone free
  • Bacterial and particle filtration to avoid contamination
  • Suitable for use in ISO 7 or higher cleanrooms
Specifications
Product Type Cleanroom Face Veil
Color White
Fastening Type Headband
Material Polyester
Size Standard
Sterility Non-sterile
Style Veil
Cleanroom Class Class 10,000 (ISO 7)
Disposable Single-use
Latex Free Yes
Length (English) 15 in.
Length (Metric) 38.1 cm
Model 62757
Packaging Type Double-bagged
Series Kimtech™ M7
Show More Show Less