missing translation for 'onlineSavingsMsg'
Learn More

Ansell Kimtech™ A5 Clean Processed Cleanroom Coveralls, Individually Packaged
GREENER_CHOICE

Catalog No. 19812856
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19812856 Small
19812857 Medium
19812858 Large
19812859 X-Large
19812860 2X-Large
19812861 3X-Large
19812862 4X-Large
18042914 6X-Large
18042915 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19812856 Supplier Ansell Supplier No. 88842

Coveralls feature a polypropylene breathable SMS fabric with a cloth-like feel.

Kimtech™ A5 Clean Processed Cleanroom Coveralls have an average bacterial filtration efficiency (BFE) of 96% and are made of 100% polypropylene breathable SMS fabric, which gives the wearer cloth-like comfort.

The Reflex Coverall Design is 7.5 times more rip-resistant than ANSI minimum standards and offers excellent fit and freedom of movement for the wearer. This personal protective apparel is triple-bagged and vacuum packaged, suitable for ISO Class 5 or higher cleanrooms.

It is made without latex, silicone and the BHT preservative and is designed for single-use (disposable PPE). The easy-to-don, individually packaged coveralls feature a mandarin collar, thumb loops and tunneled elastics.

  • 25 Kimtech™ A5 Clean Processed Cleanroom Coveralls/Case
  • Triple bagged, vacuum packed, suitable for ISO Class 5 or higher cleanrooms
  • Reflex Coverall Design offers excellent fit and freedom of movement and 7.5 times more rip-resistant than ANSI minimums
  • This protective apparel isn't made with natural rubber latex, BHT preservative or silicone
  • Kimtech™ A5 Cleanroom Apparel is manufactured in an ISO-certified facility, clean-processes and individually packaged under cleanroom conditions

Specifications

Product Type Cleanroom Coveralls
Size Small
Material Polypropylene, SMS
Color White
Disposable Single-use
Cuff Style Thumb Loop
Neck Style Collared
Seam Type Bound
Sterility Non-sterile
Collar Type Mandarin
Model Kimtech™ A5
For Use With (Application) Compounding, Mixing, Filling, Cleaning, Pharmaceutical, Biomedial Research
Packaging Individually Packaged
Cleanroom Class Class 100-100,000 (ISO 5-8)
Show More Show Less