missing translation for 'onlineSavingsMsg'
Learn More

KleenGuard™ G80 Nitrile Chemical Resistant Gloves

Green, hand-specific gloves provide maximum protection against chemical and mechanical hazards. KleenGuard™ G80 Nitrile Chemical Resistant Gloves include improved, more consistent cotton flock lining for added comfort.

Supplier:  KleenGuard™ 94447

 View more versions of this product

Catalog No. 19-812-421


Only null left
Explore available promotions

Description

Description

  • Hand specific latex-free gloves protect hands against chemical or mechanical hazards
  • Diamond embossed fingertips offer enhanced wet and dry grip
  • Flock-lined for easy donning and extra comfort in extended use
  • Improved AQL of 0.65 versus 1.5, resulting in fewer pinholes
  • Modified Nitrile formulation for more durability and enhanced chemical resistance.
  • Now available in 5 bulk-packed poly bags versus individually wrapped pairs
  • Ideal for: providing PPE category 3 protection, level 4 abrasion resistance and durability in manufacturing, transport, construction, agriculture cleaning and janitorial work, when handling chemicals, oils, grease, acids, caustics or solvents
Specifications

Specifications

FDA, ASTM D3767, ASTM D412, ASTM D5151, ASTM D6319
No
15 mil
Yes
15.7 mil
Nitrile Gloves
15 mil
EN 388:4101X
Type A
Bacteria, Fungi only
Rough
Lined
KleenGuard™ G80
33 cm
CE 0120
Green
0.38 mm
Nitrile
0.4 mm
Large
0.38 mm
No
JKLNOP
Acids, bases, Solvents and Caustics
Cotton Flock
94447
13 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions