Learn More
KleenGuard™ G60 Medium Duty Polyurethane-Coated Cut-Resistant Gloves
KleenGuard™ G60 Medium Duty Polyurethane-Coated Cut-Resistant Gloves are constructed of ultra-high molecular weight polyethylene fibers to enhance breathability, and provide ANSI Cut Level 3 protection.
$136.98 - $187.79
Specifications
ANSI Abrasion Rating ANSI 4
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A2
Hand Specific
View More Specs
Products 5
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
18-042-922
View Documents
KleenGuard™
13823
Small
Case of 12 Each for $136.98
Only null left
18-042-923
View Documents
KleenGuard™
13824
Medium
Case of 12 Each for $187.79
Only null left
18-042-924
View Documents
KleenGuard™
13825
Large
Case of 12 Each for $187.79
Only null left
18-042-925
View Documents
KleenGuard™
13826
X-Large
Case of 12 Each for $187.79
Only null left
18-042-926
View Documents
KleenGuard™
13827
2X-Large
Case of 12 Each for $187.79
Only null left
Description
Description
  • Constructed of ultra-high molecular weight polyethylene (UHMWPE) fibers to provide breathability and protection
  • Polyurethane coated palm and fingers enhance dexterity and grip capabilities
  • Separate right and left gloves in a pair provide a snug fit
Specifications
Specifications
ANSI 4
Gray
Specific
HPPE
EN 388
Knit
13
Polyurethane
ANSI A2
Yes
Cut-resistant Gloves
Assembly, plastics, light raw materials
KleenGuard™ G60
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'